Assay Details
Gene Symbol: MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040589.2 | NP_001035679.1 | 1 - 2 | 57 | 70 | |
GenBank mRNA | BC109919.1 | - | 1 - 2 | 58 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109919.1, NM_001040589.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
