Assay Details
| Gene Symbol: EEF1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040599.1 | NP_001035689.1 | 3 - 4 | 394 | 158 | |
| GenBank mRNA | BC103259.1 | - | 3 - 4 | 394 | 158 | |
More Information
Set Membership: |
|

EEF1E1
Genomic Map