Assay Details
| Gene Symbol: POLR3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046617.2 | NP_001040082.1 | 1 - 2 | 117 | 73 | |
| GenBank mRNA | BC111667.1 | - | 1 - 2 | 118 | 73 | |
More Information
Set Membership: |
|

POLR3K
Genomic Map