Assay Details
| Gene Symbol: ATP5F1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075136.2 | NP_001068604.1 | 9 - 10 | 960 | 99 | |
| GenBank mRNA | BC102466.1 | - | 9 - 10 | 961 | 99 | |
More Information
Set Membership: |
|

ATP5F1C
Genomic Map