Assay Details
Gene Symbol: EMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075324.1 | NP_001068792.1 | 2 - 3 | 176 | 84 | |
GenBank mRNA | BC111627.1 | - | 2 - 3 | 176 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
