Assay Details
| Gene Symbol: LGALS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075494.2 | NP_001068962.1 | 4 - 5 | 417 | 90 | |
| NM_001257091.1 | NP_001244020.1 | 3 - 4 | 302 | 90 | ||
| GenBank mRNA | BC120473.1 | - | 4 - 5 | 415 | 90 | |
| DT818502.1 | - | 3 - 4 | 299 | 90 | ||
| DT820728.1 | - | 3 - 4 | 303 | 90 | ||
More Information
Set Membership: |
|

LGALS2
Genomic Map