Assay Details
| Gene Symbol: UOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075648.2 | NP_001069116.1 | 1 - 2 | 85 | 85 | |
| GenBank mRNA | BC105244.1 | - | 1 - 2 | 86 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105244.1, NM_001075648.2 transcripts.
Set Membership: |
|

UOX
Genomic Map