Assay Details
| Gene Symbol: UBA52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076363.2 | NP_001069831.1 | 1 - 2 | 30 | 64 | |
| GenBank mRNA | BC102248.1 | - | 1 - 2 | 32 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102248.1, NM_001076363.2 transcripts.
Set Membership: |
|

UBA52
Genomic Map