Assay Details
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076473.2 | NP_001069941.1 | 5 - 6 | 1323 | 85 | |
GenBank mRNA | BC102920.1 | - | 5 - 6 | 1324 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
