Assay Details
| Gene Symbol: WFDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076490.2 | NP_001069958.1 | 1 - 2 | 98 | 76 | |
| GenBank mRNA | BC102201.1 | - | 1 - 2 | 99 | 76 | |
More Information
Set Membership: |
|

WFDC2
Genomic Map