Assay Details
| Gene Symbol: RPL13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076998.2 | NP_001070466.1 | 4 - 5 | 274 | 61 | |
| GenBank mRNA | BC103039.1 | - | 4 - 5 | 275 | 61 | |
| BC105558.1 | - | 2 - 3 | 1060 | 61 | ||
More Information
Set Membership: |
|

RPL13A
Genomic Map