Assay Details
| Gene Symbol: RBBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077013.2 | NP_001070481.1 | 10 - 11 | 1107 | 68 | |
| GenBank mRNA | BC105195.1 | - | 10 - 11 | 1108 | 68 | |
| BT030530.1 | - | 10 - 11 | 1124 | 68 | ||
More Information
Set Membership: |
|

RBBP4
Genomic Map