Assay Details
| Gene Symbol: NDUFAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077029.2 | NP_001070497.1 | 1 - 2 | 167 | 151 | |
| GenBank mRNA | BC108241.1 | - | 1 - 2 | 168 | 151 | |
More Information
Set Membership: |
|

NDUFAF2
Genomic Map