Assay Details
Gene Symbol: RIIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077049.1 | NP_001070517.1 | 2 - 3 | 175 | 95 | |
GenBank mRNA | BC108187.1 | - | 2 - 3 | 175 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
