Assay Details
| Gene Symbol: C16H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077054.1 | NP_001070522.1 | 257 | 109 | ||
| GenBank mRNA | BC109731.1 | - | 257 | 109 | ||
More Information
Set Membership: |
|

C16H1orf100
Genomic Map