Assay Details
| Gene Symbol: C22H3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077085.2 | NP_001070553.1 | 1 - 2 | 112 | 68 | |
| GenBank mRNA | BC111666.1 | - | 1 - 2 | 113 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111666.1, NM_001077085.2 transcripts.
Set Membership: |
|

C22H3orf84
Genomic Map