Assay Details
| Gene Symbol: C22H3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077085.2 | NP_001070553.1 | 3 - 4 | 291 | 72 | |
| GenBank mRNA | BC111666.1 | - | 3 - 4 | 292 | 72 | |
More Information
Set Membership: |
|

C22H3orf84
Genomic Map