Assay Details
| Gene Symbol: INSL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077521.2 | NP_001070989.1 | 1 - 2 | 362 | 84 | |
| GenBank mRNA | BC109721.1 | - | 1 - 2 | 363 | 84 | |
More Information
Set Membership: |
|

INSL6
Genomic Map