Assay Details
Gene Symbol: CYCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077963.2 | NP_001071431.1 | 1 - 2 | 109 | 73 | |
GenBank mRNA | BC102714.1 | - | 1 - 2 | 110 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102714.1, NM_001077963.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
