Assay Details
| Gene Symbol: CYBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078087.2 | NP_001071555.1 | 4 - 5 | 266 | 58 | |
| GenBank mRNA | BC102788.1 | - | 4 - 5 | 267 | 58 | |
More Information
Set Membership: |
|

CYBC1
Genomic Map