Assay Details
Gene Symbol: CDC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078153.2 | NP_001071621.1 | 1 - 2 | 162 | 77 | |
GenBank mRNA | BC102716.1 | - | 1 - 2 | 163 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102716.1, NM_001078153.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
