Assay Details
| Gene Symbol: RSL24D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080355.2 | NP_001073824.1 | 5 - 6 | 458 | 73 | |
| GenBank mRNA | BC103265.1 | - | 5 - 6 | 460 | 73 | |
More Information
Set Membership: |
|

RSL24D1
Genomic Map