Assay Details
Gene Symbol: DSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083785.1 | NP_001077254.1 | 3 - 4 | 652 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
