Assay Details
| Gene Symbol: ASS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173892.4 | NP_776317.1 | 1 - 2 | 40 | 49 | |
| GenBank mRNA | BC102474.1 | - | 1 - 2 | 41 | 49 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102474.1, NM_173892.4 transcripts.
Set Membership: |
|

ASS1
Genomic Map