Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173929.3 | NP_776354.2 | 6 - 7 | 604 | 115 | |
| GenBank mRNA | BC108213.1 | - | 6 - 7 | 604 | 115 | |
| X14712.1 | - | 609 | 115 | |||
More Information
Set Membership: |
|

PAEP
Genomic Map