Assay Details
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173935.3 | NP_776360.2 | 2 - 3 | 131 | 69 | |
GenBank mRNA | AB036426.1 | - | 1 - 2 | 83 | 69 | |
BC103211.1 | - | 2 - 3 | 132 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
