Assay Details
| Gene Symbol: S100A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174651.3 | NP_777076.1 | 1 - 2 | 412 | 96 | |
| GenBank mRNA | AF011757.1 | - | 1 - 2 | 137 | 96 | |
| BC102542.1 | - | 1 - 2 | 413 | 96 | ||
| D49548.1 | - | 2 - 3 | 184 | 96 | ||
| FJ358602.1 | - | 2 - 3 | 206 | 96 | ||
More Information
Set Membership: |
|

S100A12
Genomic Map