Assay Details
| Gene Symbol: ATP5PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174724.4 | NP_777149.1 | 43 | 97 | ||
| GenBank mRNA | BC104564.1 | - | 44 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC104564.1, NM_174724.4 transcripts.
Set Membership: |
|

ATP5PD
Genomic Map