Assay Details
| Gene Symbol: ATP5PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174724.4 | NP_777149.1 | 351 | 77 | ||
| GenBank mRNA | BC104564.1 | - | 352 | 77 | ||
| X06089.1 | - | 301 | 77 | |||
More Information
Set Membership: |
|

ATP5PD
Genomic Map