Assay Details
| Gene Symbol: RAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175824.2 | NP_787018.1 | 7 - 8 | 781 | 92 | |
| GenBank mRNA | BC102330.1 | - | 7 - 8 | 781 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAP1B| Gene Symbol: RAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175824.2 | NP_787018.1 | 7 - 8 | 781 | 92 | |
| GenBank mRNA | BC102330.1 | - | 7 - 8 | 781 | 92 | |
Set Membership: |
|