Assay Details
| Gene Symbol: COX7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY911320.1 | - | 1 - 2 | 175 | 104 | |
| BC102263.1 | - | 1 - 2 | 172 | 104 | ||
| X15725.1 | - | 133 | 104 | |||
More Information
Set Membership: |
|

COX7C
Genomic Map