Assay Details
| Gene Symbol: IDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181012.3 | NP_851355.2 | 5 - 6 | 729 | 87 | |
| GenBank mRNA | AF136009.1 | - | 4 - 5 | 609 | 87 | |
| BC103368.1 | - | 5 - 6 | 729 | 87 | ||
| DQ403090.1 | - | 3 - 4 | 432 | 87 | ||
More Information
Set Membership: |
|

IDH1
Genomic Map