Assay Details
| Gene Symbol: APOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001175.2 | NP_001001175.1 | 37 | 83 | ||
| GenBank mRNA | BC111661.1 | - | 37 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111661.1, NM_001001175.2 transcripts.
Set Membership: |
|

APOC3
Genomic Map