Assay Details
| Gene Symbol: APOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001175.2 | NP_001001175.1 | 100 | 87 | ||
| GenBank mRNA | AB126687.1 | - | 1 - 2 | 80 | 87 | |
| BC111661.1 | - | 100 | 87 | |||
More Information
Set Membership: |
|

APOC3
Genomic Map