Assay Details
| Gene Symbol: DMAC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007812.3 | NP_001007813.1 | 4 - 5 | 548 | 127 | |
| GenBank mRNA | AY780292.1 | - | 4 - 5 | 537 | 127 | |
| BC109757.1 | - | 4 - 5 | 550 | 127 | ||
More Information
Set Membership: |
|

DMAC2L
Genomic Map