Assay Details
| Gene Symbol: SPAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008413.3 | NP_001008413.3 | 1 - 2 | 113 | 147 | |
| GenBank mRNA | BC110183.1 | - | 1 - 2 | 111 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC110183.1, NM_001008413.3 transcripts.
Set Membership: |
|

SPAM1
Genomic Map