Assay Details
| Gene Symbol: MBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010994.3 | NP_001010994.1 | 4 - 5 | 530 | 80 | |
| GenBank mRNA | AB178774.1 | - | 4 - 5 | 450 | 80 | |
| AY861663.1 | - | 3 - 4 | 372 | 80 | ||
| BC109674.1 | - | 4 - 5 | 531 | 80 | ||
More Information
Set Membership: |
|

MBL1
Genomic Map