Assay Details
| Gene Symbol: RPL35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025321.3 | NP_001020492.1 | 231 | 114 | ||
| GenBank mRNA | AY911347.1 | - | 3 - 4 | 233 | 114 | |
| BC111654.1 | - | 232 | 114 | |||
More Information
Set Membership: |
|

RPL35A
Genomic Map