Assay Details
Gene Symbol: MARCHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037589.1 | NP_001032678.1 | 1 - 2 | 101 | 127 | |
GenBank mRNA | BC109744.1 | - | 1 - 2 | 101 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109744.1, NM_001037589.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
