Assay Details
Gene Symbol: MARCHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037589.1 | NP_001032678.1 | 3 - 4 | 396 | 67 | |
GenBank mRNA | BC109744.1 | - | 3 - 4 | 396 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
