Assay Details
| Gene Symbol: GRINA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037593.1 | NP_001032682.1 | 5 - 6 | 911 | 60 | |
| GenBank mRNA | BC109761.1 | - | 5 - 6 | 911 | 60 | |
More Information
Set Membership: |
|

GRINA
Genomic Map