Assay Details
| Gene Symbol: GLRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037604.2 | NP_001032693.1 | 1 - 2 | 291 | 92 | |
| GenBank mRNA | BC109978.1 | - | 1 - 2 | 294 | 92 | |
More Information
Set Membership: |
|

GLRX
Genomic Map