Assay Details
Gene Symbol: POLR2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037623.3 | NP_001032712.1 | 1 - 2 | 73 | 61 | |
GenBank mRNA | BC103450.1 | - | 1 - 2 | 47 | 61 | |
EE222335.1 | - | 1 - 2 | 74 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103450.1, NM_001037623.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
