Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038023.2 | NP_001033112.1 | 2 - 3 | 174 | 62 | |
| GenBank mRNA | BC109575.1 | - | 2 - 3 | 175 | 62 | |
More Information
Set Membership: |
|

NIT1
Genomic Map