Assay Details
| Gene Symbol: AURKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038028.1 | NP_001033117.1 | 3 - 4 | 583 | 76 | |
| GenBank mRNA | BC111181.1 | - | 3 - 4 | 583 | 76 | |
| DQ334808.1 | - | 3 - 4 | 377 | 76 | ||
More Information
Set Membership: |
|

AURKA
Genomic Map