Assay Details
Gene Symbol: UBE2T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038034.2 | NP_001033123.1 | 3 - 4 | 267 | 68 | |
GenBank mRNA | BC109637.1 | - | 3 - 4 | 268 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
