Assay Details
| Gene Symbol: TRMT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038040.2 | NP_001033129.1 | 8 - 9 | 1352 | 62 | |
| GenBank mRNA | BC111249.1 | - | 8 - 9 | 1354 | 62 | |
More Information
Set Membership: |
|

TRMT6
Genomic Map