Assay Details
| Gene Symbol: PRR13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038047.1 | NP_001033136.1 | 1 - 2 | 90 | 77 | |
| GenBank mRNA | BC111340.1 | - | 1 - 2 | 90 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111340.1, NM_001038047.1 transcripts.
Set Membership: |
|

PRR13
Genomic Map