Assay Details
| Gene Symbol: NUP50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038048.2 | NP_001033137.1 | 4 - 5 | 774 | 60 | |
| GenBank mRNA | BC109804.1 | - | 4 - 5 | 775 | 60 | |
More Information
Set Membership: |
|

NUP50
Genomic Map