Assay Details
| Gene Symbol: KCTD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038057.2 | NP_001033146.1 | 1 - 2 | 404 | 78 | |
| GenBank mRNA | BC111242.1 | - | 1 - 2 | 405 | 78 | |
More Information
Set Membership: |
|

KCTD13
Genomic Map